missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MC2R Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518867
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
MC2R encodes one member of the five-member G-protein associated melanocortin receptor family. Melanocortins are peptides derived from pro-opiomelanocortin. MC2R is selectively activated by adrenocorticotropic hormone, whereas the other four melanocortin receptors recognize a variety of melanocortin ligands. Mutations in MC2R can result in familial glucocorticoid deficiency.Specifications
MC2R | |
Polyclonal | |
Unconjugated | |
MC2R | |
ACTH receptor; ACTHR; ACTH-R; Adrenocorticotropic hormone receptor; adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2R; MC2-R; melanocortin 2 receptor; melanocortin 2 receptor (adrenocorticotropic hormone); melanocortin receptor 2; MGC125798 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4158 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q01718 | |
MC2R | |
Synthetic peptide sequence (ENINNTARNNSDC) corresponding to the N-terminus amino acids of MC2R | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |