missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIME Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA19064
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
LIME Polyclonal antibody specifically detects LIME in Mouse samples. It is validated for Western Blot.Specifications
LIME | |
Polyclonal | |
Unconjugated | |
Lime1 | |
2810038K19Rik; dJ583P15.4; FLJ20406; Lck interacting transmembrane adaptor 1; lck-interacting membrane protein; Lck-interacting molecule; lck-interacting transmembrane adapter 1; Lime; lime 1; LIME1; LP8067; RP4-583P15.5 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
72699 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9EQR5 | |
Lime1 | |
Synthetic peptide corresponding to amino acids ENVYESIKEMGL. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |