Learn More
ITGB6 Monoclonal Antibody (437211), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA523675
Product Code. 15583362
  • £440.00 / 100 tests
Explore more special offers
Description
Description
ITGB6 Monoclonal antibody specifically detects ITGB6 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ITGB6
Monoclonal
APC
Itgb6
2210409C20Rik; 4831415H04Rik; AI1H; integrin beta 6; integrin beta-6; integrin subunit beta 6; integrin, beta 6; Itgb6
Mouse
Protein A/G
RUO
3694
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
437211
Buffered saline solution with BSA and 0.09% sodium azide
P18564
Itgb6
CHO-derived recombinant human Integrin alpha V beta 6 Phe31-Val992 (Integrin alpha V) and Gly22-Asn707 (Integrin beta 6)
100 Tests
Primary
Human
Antibody
IgG2b