missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG2b, kappa, BB515, Clone: R35-38, BD
Brand: BD Biosciences 564421
330.37 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone: R35-38
Isotype: IgG2b, κ
Flow Cytometry, Isotype control
Specifications
IgG2b κ | |
Brilliant Blue 515 (BB515) | |
Affinity Purified | |
IgG2b κ |
R35-38 | |
Rat | |
0.1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction