missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2a κ, Mouse, V450, Clone: MOPC-173, BD

Brand:  BD Biosciences 560882

289.48 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15890559

  • £293.00 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Intracellular Staining, Isotype Control

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot
Violet Fluor 450 (V450)
Affinity Purified
4°C
MOPC-173
Mouse
0.1 mg
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.