Learn More
anti-IgG1 κ Mouse, FITC, Clone: MOPC-21, Isotype Control, BD
For low background screening on a variety of mouse and human tissues. anti-IgG1 κ Mouse, FITC, Clone: MOPC-21, Isotype Control, BD immunoglobulin is a mouse myeloma protein. This antibody is routinely tested by flow cytometric analysis.
Brand: BD Biosciences 555748
Additional Details : Weight : 0.80000kg
Description
- Host Species: Mouse
- Clone: MOPC-21
- Isotype: IgG1, κ
- Applications: Flow Cytometry, Isotype Control
- Other applications were tested at BD Biosciences Pharmingen during antibody development only
Specifications
Flow Cytometry | |
MOPC-21 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
IgG1 κ |
Monoclonal | |
FITC | |
Mouse | |
100 Tests | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |