missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA9 Monoclonal Antibody (9F8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1094
Code : PN
Additional Details : Weight : 0.01000kg
Description
MA1-094 has been successfully used in Western blot, immunofluorescence, and immunoprecipitation applications on human, monkey, and rat samples.
The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.Specifications
HSPA9 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P38646, P48721 | |
HSPA9 | |
Protein expressed in 293T cell transfected with human HSPA9 expression vector | |
100 μg | |
Primary | |
Human, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
9F8 | |
Unconjugated | |
HSPA9 | |
70 kDa heat shock protein; 74kDa; 75 kDa glucose-regulated protein; C3H-specific antigen; catecholamine-regulated protein 40; CRP40; CSA; epididymis secretory sperm binding protein Li 124m; EVPLS; GRP 75; GRP75; GRP-75; heat shock 70 kDa protein 9; heat shock 70 kDa protein 9B; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; heat shock 70kDa protein 9B (mortalin-2); Heat shock protein; heat shock protein 9; heat shock protein 9A; heat shock protein cognate 74; heat shock protein family A (Hsp70) member 9; heat shock protein family A (Hsp70) member 9 S homeolog; heat shock protein, 74 kDa, A; heat shock protein, A; HEL-S-124m; Hsc74; HSP; Hsp74; Hsp74a; hspa9; hspa9.S; Hspa9a; hspa9-a; hspa9b; hspa9-b; hypothetical protein; I79_009139; MGC4500; mortalin; mortalin, perinuclear; mortalin2; mortalin-2; mot; MOT2; Mot-2; Mthsp70; mt-HSP70; MTHSP75; p66 MOT; p66-mortalin; pbp74; Peptide-binding protein 74; RCJMB04_2m8; SAAN; SIDBA4; stress-70 p | |
Mouse | |
Protein A | |
RUO | |
291671, 3313 | |
-20°C | |
Liquid |