Learn More
HSF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530064
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, mouse liver. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (81%), Chicken (87%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.Specifications
HSF2 | |
Polyclonal | |
Unconjugated | |
HSF2 | |
AI661205; heat shock factor 2; heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; Hsf2; HSTF 2; HSTF2; MGC117376; MGC156196; MGC75048 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15500, 3298 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P38533, Q03933 | |
HSF2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 188 of Human HSF2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |