Learn More
homeobox C8, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human HOXC8 protein.
Brand: Abnova H00003224-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders. [provided by RefSeq
Sequence: MSSYFVNPLFSKYKAGESLEPAYYDCRFPQSVGRSHALVYGPGGSAPGFQHASHHVQDFFHHGTSGISNSGYQQNPCSLSCHGDASKFYGYEALPRQSLYGAQQEASVVQYPDCKSSANTNSSEGQGHLNQNSSPSLMFPWMRPHAPGRRSGRQTYSRYQTLELEKEFLFNPYLTRKRRIEVSHALGLTERQVKIWFQNRRMKWKKENNKDKLPGARDEEKVEEEGNEEEEKEEEEKEENKDSpecifications
homeobox C8 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human HOXC8 protein. | |
HOXC8 | |
HOX3/HOX3A | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022658.3 | |
HOXC8 | |
HOXC8 (NP_073149.1, 1 a.a. ∼ 242 a.a) full-length human protein. | |
50 μg | |
Yes | |
3224 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |