missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2 (HER-2) Monoclonal Antibody (e2-4001), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen AHO1011
This item is not returnable.
View return policy
Description
ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
Specifications
ErbB2 (HER-2) | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P04626, P70424 | |
ERBB2 | |
Cytoplasmic domain of human c-erbB-2 protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
e2-4001 | |
Unconjugated | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb | |
Mouse | |
Protein G | |
RUO | |
13866, 2064 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ErbB2 (HER-2) Monoclonal Antibody (e2-4001), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction