missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518187
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene.Specifications
FBXO44 | |
Polyclonal | |
Unconjugated | |
FBXO44 | |
5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
93611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9H4M3 | |
FBXO44 | |
Synthetic peptide sequence (AALTPPEPPSAEP) corresponding to the C-terminus amino acids of FBXO44 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |