Learn More
FASTK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528671
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4. Predicted reactivity: Mouse (91%), Dog (96%), Pig (92%), Chimpanzee (99%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase was shown to be activated rapidly during Fas-mediated apoptosis in Jurkat cells. In response to Fas receptor ligation, it phosphorylates TIA1, an apoptosis-promoting nuclear RNA-binding protein. The encoded protein is a strong inducer of lymphocyte apoptosis. Two transcript variants encoding different isoforms have been found for this gene. Other variants exist, but their full-length natures have not yet been determined.Specifications
FASTK | |
Polyclonal | |
Unconjugated | |
FASTK | |
0610011K02Rik; 610011K02Rik; Fas activated serine/threonine kinase; Fas-activated serine/threonine kinase; FAST; FAST kinase; Fastk | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.88 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q14296 | |
FASTK | |
Recombinant fragment corresponding to a region within amino acids 306 and 549 of Human FAST | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |