Learn More
FAM55D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542687
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TYSVKEMEYL TRAIDRTGGE KNTVIVISLG QHFRPFPIDV FIRRALNVHK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
FAM55D Polyclonal antibody specifically detects FAM55D in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FAM55D | |
Polyclonal | |
Unconjugated | |
NXPE4 | |
C11orf33; C130036J11; D930028F11Rik; Fam55b; Fam55d; family with sequence similarity 55, member B; family with sequence similarity 55, member D; hypothetical protein LOC244853; hypothetical protein LOC515648; LOC100731209; neurexophilin and PC-esterase domain family member 4; neurexophilin and PC-esterase domain family, member 4; NXPE family member 4; Nxpe4; Protein FAM55D; UNQ3018/PRO9799 | |
Rabbit | |
Protein A | |
RUO | |
54827 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UWF7 | |
NXPE4 | |
Synthetic peptide directed towards the C-terminal of human FAM55D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |