Learn More
Factor XI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521713
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Dog (83%), Rabbit (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality.Specifications
Factor XI | |
Polyclonal | |
Unconjugated | |
F11 | |
1600027G01Rik; AI503996; Cf11; Coagulation factor XI; Coagulation factor XIa heavy chain; Coagulation factor XIa light chain; F11; FXI; LRRGT00086; plasma thromboplastin antecedent; PTA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P03951 | |
F11 | |
Recombinant fragment corresponding to a region within amino acids 1 and 312 of Factor XI (Uniprot ID#P03951) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |