missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXTL1 Monoclonal Antibody (353713), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524098
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) EXTL1, rhEXTL2, rhEXTL3, or recombinant mouse EXTL2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
EXTL1 Monoclonal antibody specifically detects EXTL1 in Mouse samples. It is validated for Immunoprecipitation, Western Blot.Specifications
EXTL1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9JKV7 | |
Extl1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 1/EXTL1 Ala30-Pro669 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
353713 | |
Unconjugated | |
Extl1 | |
alpha-N-acetylglucosaminyltransferase II; D430033M16Rik; EC 2.4.1.224; exostoses (multiple)-like 1; exostoses-like 1; exostosin like glycosyltransferase 1; exostosin-L; Exostosin-like 1; Exostosin-like 1 (Exostosin-L) (Multiple exostosis-like protein); exostosin-like glycosyltransferase 1; EXTL; Extl1; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase; glucuronyl-N-acetylglucosaminylproteoglycan alpha-1,4-N- acetylglucosaminyltransferase; MGC70794; multiple exostoses-like 1; multiple exostosis-like protein | |
Rat | |
Protein A/G | |
RUO | |
56219 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |