missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM62A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532764
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Orangutan, Gibbon, Monkey (94%) Marmoset (88%) Bovine, Dog, Horse, Pig (81%).
May play a role as calcium-regulated intrinsic membrane protein. Target protein is an enriched in adipocyte plasma membranes and high density microsome fractions.Specifications
FAM62A | |
Polyclonal | |
Unconjugated | |
ESYT1 | |
ESYT1; E-Syt1; extended synaptotagmin 1; extended synaptotagmin like protein 1; extended synaptotagmin protein 1; extended synaptotagmin-1; extended synaptotagmin-like protein 1; FAM62A; family with sequence similarity 62 (C2 domain containing), member A; KIAA0747; MBC2; membrane bound C2 domain containing protein; membrane-bound C2 domain-containing protein; vp115 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23344 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BSJ8 | |
ESYT1 | |
Synthetic 16 amino acid peptide from C-terminus of human ESYT1 / FAM62A. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |