missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119 Monoclonal Antibody (TER119), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517821
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.Specifications
TER-119 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
0 | |
Ly76 | |
Day 14 BALB/c fetal liver cells | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry | |
TER119 | |
FITC | |
Ly76 | |
Ly76; Ly-76; TER119 | |
Rat | |
Protein G | |
RUO | |
104231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |