Learn More
ERP29 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519034
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
ERP29 | |
Polyclonal | |
Unconjugated | |
Erp29 | |
1200015M03Rik; 2810446M09Rik; AW209030; C12orf8; endoplasmic reticulum lumenal protein ERp28; endoplasmic reticulum protein 29; endoplasmic reticulum protein ERp29; endoplasmic reticulum resident protein 28; Endoplasmic reticulum resident protein 29; endoplasmic reticulum resident protein 31; endoplasmic retuclum protein 29; epididymis secretory protein Li 107; Erp28; ERp29; Erp31; HEL-S-107; PDIA9; PDI-DB; protein disulfide isomerase family A, member 9 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10961, 117030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P30040, P52555 | |
Erp29 | |
Synthetic peptide sequence (EKNKMSDGKKEELQ) corresponding to the C-terminus amino acids of ERP29 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |