Learn More
DYX1C1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531138
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, H1299, MCF-7. Predicted reactivity: Human (99%), Mouse (87%), Rat (88%), Xenopus laevis (80%), Chicken (82%), Chimpanzee (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DYX1C1 Polyclonal antibody specifically detects DYX1C1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
DYX1C1 | |
Polyclonal | |
Unconjugated | |
DNAAF4 | |
1700010I24Rik; b2b811Clo; CILD25; DNAAF4; Dynein assembly factor 4, axonemal; dynein, axonemal, assembly factor 4; dyslexia susceptibility 1 candidate 1; dyslexia susceptibility 1 candidate 1 homolog; dyslexia susceptibility 1 candidate 1 homolog (human); dyslexia susceptibility 1 candidate gene 1 protein; Dyslexia susceptibility 1 candidate gene 1 protein homolog; DYX1; Dyx1c1; DYXC1; Edem2; Ekn1; FLJ37882; MGC70618; RD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
161582 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8WXU2 | |
DNAAF4 | |
Recombinant protein fragment corresponding to a region within amino acids 183 and 406 of Human DYX1C1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |