Learn More
Factor VII Monoclonal Antibody (406722), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524143
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) FGFA, rhFactor VII, rhFactor X, rhFactor XI, rhPROC, rhThrombin, rhUPA1, rhKLK-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, or -B1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Factor VII Monoclonal antibody specifically detects Factor VII in Mouse samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Factor VII | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P70375 | |
F7 | |
CHO-derived recombinant mouse Coagulation Factor VII Ala42-Leu446 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
406722 | |
Unconjugated | |
F7 | |
AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
Rat | |
Protein A/G | |
RUO | |
14068 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |