missing translation for 'onlineSavingsMsg'
Learn More

CD20, Mouse anti-Human, Clone: 2H7, BUV395, BD

Brand:  BD Biosciences 563781

165.21 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15843089

  • £168.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: 2H7
Isotype: IgG2b, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human 6.16c1.3 B cell line

Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD20
2H7
Mouse
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
Monoclonal
Brilliant Ultraviolet 395
Human 6.16c1.3 B cell line
25 Tests
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.