missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APIP Monoclonal Antibody (19F461), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA141247
Code : LE
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: HeLa or 293, antigen standard for APIP (transient overexpression lysate).
APIP Monoclonal antibody specifically detects APIP in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
APIP | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q96GX9, Q9WVQ5 | |
APIP | |
Recombinant APIP protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot | |
19F461 | |
Unconjugated | |
APIP | |
APAF1 interacting protein; APAF1-interacting protein; APIP; APIP2; CG129; CGI29; CGI-29; dJ179L10.2; hAPIP; Methylthioribulose-1-phosphate dehydratase; Mmrp19; monocyte macrophage 19; monocyte/macrophage protein 19; MTRu-1-P dehydratase; probable methylthioribulose-1-phosphate dehydratase; RGD1564562 | |
Mouse | |
Protein G | |
RUO | |
51074, 56369 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |