missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiphysin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518513
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and rat based on sequence homology.
Amphiphysin Polyclonal antibody specifically detects Amphiphysin in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Amphiphysin | |
Polyclonal | |
Unconjugated | |
Amph | |
Amph; AMPH1; amphiphysin; amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen); amphiphysin 1; amphiphysin I; HGNC:471 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
218038, 273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P49418, Q7TQF7 | |
Amph | |
Synthetic peptide sequence (KRKDESRISKAE) corresponding to the internal amino acids of AMPH | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |