Learn More
AKR7A2 Monoclonal Antibody (2H3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA520447
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MSRPPPPRVA SVLGTMEMGR RMDAPASAAA VRAFLERGHT ELDTAFMYSD GQSETILGGL GLGLGGGDCR VKIATKANPW DGKSLKPDSV RSQLETSLKR LQCPQVDLFY LHAPDHGTPV EETLHACQRL HQEGKFVELG LSNYASWEVA EICTLCKSNG WILPTVYQGM YNATTRQVET ELFPCLRHFG LRFYAYNPLA GGLLTGKYKY EDKDGKQPVG RFFGNSWAET YRNRFWKEHH FEAIALVEKA LQAAYGASAP SVTSAALRWM YHHSQLQGAH GDAVILGMSS LEQLEQNLAA TEEGPLEPAV VDAFNQAWHL VAHECPNYFR
AKR7A2 Monoclonal antibody specifically detects AKR7A2 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Specifications
AKR7A2 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
O43488 | |
AKR7A2 | |
AKR7A2 (AAH10852.1, 1 a.a. ∽ 330 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunoprecipitation, Western Blot | |
2H3 | |
Unconjugated | |
AKR7A2 | |
0610025K21Rik; Afar; AFAR1; Afar2; AFB1 aldehyde reductase 1; AFB1-AR 1; AFB1-AR1; aflatoxin aldehyde reductase; aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; Aiar; AKR7; AKR7A2; Akr7a5; aldo-keto reductase family 7 member A2; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase); aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; rAFAR2; SSA reductase; succinic semialdehyde reductase; synonyms: AFAR1 | |
Mouse | |
Affinity chromatography | |
RUO | |
8574 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |