missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1CL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539013
Code : NEW
Additional Details : Weight : 0.00900kg
Specifications
AKR1CL1 | |
Polyclonal | |
Unconjugated | |
AKR1C8P | |
AKR1C8P | |
A synthetic peptide derived from the internal region of human AKR1CL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5T2L2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340811 | |
-20°C | |
Liquid |