Learn More
IBA1 Monoclonal Antibody (1022-5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA140227
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-40227 detects AIF1 from human and rat samples. MA1-40227 has been successfully used in ELISA, immunohistochemistry (paraffin) and Western blot applications. The MA1-40227 immunogen is human AIF1 polypeptide.
AIF1 is induced by cytokines and interferon. It is thought to be involved in the negative regulation of growth of vascular smooth muscle cells, which contributes to the anti-inflammatory response to vessel wall trauma.Specifications
IBA1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P55008, P55009 | |
AIF1 | |
Recombinant human AIF-1 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1022-5 | |
Unconjugated | |
AIF1 | |
AI607846; AIF1; AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; Bart1; BART-1; D17H6S50E; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; G1; Iba; Iba1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; ionized calcium-binding adapter molecule 1; IRT1; IRT-1; microglia response factor; Mrf1; MRF-1; protein BART-1; protein G1; testis specific | |
Mouse | |
Protein G | |
RUO | |
199, 29427 | |
4° C | |
Liquid |