Learn More
Adiponectin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129901
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-29901 detects Adiponectin from rat, mouse, human samples. PA1-29901 has been successfully used in immunoprecipitation, and Western blot applications. The PA1-29901 immunogen is: E.coli expressed recombit full length protein (Rat). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Adipose cells produce and secrete numerous physiologically important proteins, such as Lipoprotein Lipase, Leptin, and Adipocyte Complement Related protein of 30 kDa, also know as Acrp30 or Adiponectin. Adiponectin is a circulating protein that is secreted exclusively by differentiated adipocytes. During adipocyte differentiation, Adiponectin mRNA is induced > 100 fold. Adiponmectin imporves the ability of insulin to suppress glucose production, at sub physiological levels, thereby linking adipose tissue to whole body glucose regulation. Adiponectin function appears to be regulated by phosphatidylinositol 3 kinase (PI3k) since Adiponectin secretion is blocked by pharmacologic inhibitors of this kinase. Adiponectin mRNA is signigicantly reduced in adipose tissue of obese patients with type 2 diabetes. The structural similarity of Adiponectin to TNF alpha suggests that Adiponectin may play a role in pathogenesis of insulin resistance in Type 2 diabetes. Adiponectin is implicated as a regulator of whole body energy homeostasis.Specifications
Adiponectin | |
Polyclonal | |
Unconjugated | |
Q15848, Q60994 | |
ADIPOQ | |
E.coli expressed recombit full length protein (Rat). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
ADIPOQ | |
Adipocyte, C1q and collagen domain-containing protein, 30 kDa adipocyte complement-related protein, ACRP30, Adipose most abundant gene transcript 1, a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11450, 246253, 9370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |