Learn More
Adenylate Kinase 7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527927
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (87%), Rat (88%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The adenylate kinases (AK) are a family of structurally and functionally related enzymes that catalyze a similar reaction, MgNTP + AMP = MgNDP + ADP (N = A or G). The AK enzymes are important for maintenance of homeostasis of the adenine and guanine nucleotide pools. AK1 is a cytosolic enzyme for which ATP is the substrate. AK2 catalyzes the same reaction as AK1, but it is localized in the mitochondrial intermembrane space. AK3 is present in the mitochondrial matrix and prefers GTP over ATP as the substrate.Specifications
Adenylate Kinase 7 | |
Polyclonal | |
Unconjugated | |
AK7 | |
4930502N02Rik; adenylate kinase 7; AK 7; AK7; ATP-AMP transphosphorylase 7; putative adenylate kinase 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
122481 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96M32 | |
AK7 | |
Recombinant fragment corresponding to a region within amino acids 205 and 706 of Human Adenylate kinase 7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |