Learn More
Adenovirus Hexon Monoclonal Antibody (E28H), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17330
Code : LD
Additional Details : Weight : 0.01000kg
Description
The MA1-7330 antibody reacts with Adenovirus hexon in viral and infected tissue samples. MA1-7330 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7330 antibody was raised against Adenovirus hexon protein.
Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.Specifications
Adenovirus Hexon | |
Monoclonal | |
0.1 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
E28H | |
FITC | |
Adeno Virus; AdV; HADV; HAdVC_gp08 | |
Adenovirus hexon protein. | |
100 μg | |
Primary | |
4° C, store in dark | |
Liquid |