Learn More
ADORA2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1042
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-042 detects adenosine receptor A2a (A2aAR) in human, rat, mouse and canine tissues. This antibody does not detect other AR subtypes. PA1-042 has been successfully used in Western blot, ICC/IF, immunohistochemistry and immunoprecipitation procedures. By Western blot, this antibody detects an ∽45 kDa protein from canine striatum representing A2aAR. Immunohistochemical staining of A2aAR in human hippocampus with PA1-042 yields a pattern consistent with plasma membrane staining. The PA1-042 immunogen is a synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR.
ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
ADORA2A | |
Polyclonal | |
Unconjugated | |
ADORA2A | |
A2a receptor; A2AAR; A2aR; AA2AR; ADENO; adenosine A2a receptor; Adenosine A2a-receptor; adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2; ADORA2A; Adora2l1; AR; hA2aR; RDC8; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11540, 135, 25369, 403960 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.69 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P11617, P29274, P30543, Q60613 | |
ADORA2A | |
Synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR. | |
100 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG |