missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS15 Monoclonal Antibody (561819), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524252
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
ADAMTS15 Monoclonal antibody specifically detects ADAMTS15 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ADAMTS15 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8TE58 | |
ADAMTS15 | |
CHO-derived recombinant human ADAMTS15 Gly18-Cys682 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
561819 | |
Unconjugated | |
ADAMTS15 | |
a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAM metallopeptidase with thrombospondin type 1 motif, 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domains | |
Mouse | |
Protein A/G | |
RUO | |
170689 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |