missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544080
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PTETLGVENK GYFGDEQQIR TEPILPEIHF LNKPASKDSR GIADPNQSAK Sequence homology: Dog: 86%; Horse: 85%; Human: 100%; Mouse: 91%
ADAM7 Polyclonal antibody specifically detects ADAM7 in Human samples. It is validated for Western Blot.Specifications
ADAM7 | |
Polyclonal | |
Unconjugated | |
ADAM7 | |
9230104J12Rik; a disintegrin and metallopeptidase domain 7; a disintegrin and metalloprotease domain (ADAM) 7; a disintegrin and metalloprotease domain 7; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 7; ADAM; ADAM 7; ADAM metallopeptidase domain 7; Adam7; ADAM-7; ADAMs; Disintegrin and metalloproteinase domain-containing protein 7; EAP1; EAPI; epididymal apical protein I; GP83; GP-83; metalloendopeptidases; Sperm maturation-related glycoprotein GP-83 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8756 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H2U9 | |
ADAM7 | |
Synthetic peptide directed towards the C-terminal of human ADAM7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |