missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKMY2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550324
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total ANKMY2 protein.
ANKMY2 Polyclonal antibody specifically detects ANKMY2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ANKMY2 | |
Polyclonal | |
Unconjugated | |
Ankmy2 | |
AI035571; ANKMY2; ankyrin repeat and MYND domain containing 2; ankyrin repeat and MYND domain-containing protein 2; EGK_13887; Gna14; guanine nucleotide binding protein, alpha 14; LOW QUALITY PROTEIN: ankyrin repeat and MYND domain-containing protein 2; ZMYND20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57037 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IV38 | |
Ankmy2 | |
Fusion protein corresponding to residues near the N terminal of human ankyrin repeat and MYND domain containing 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |