missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15710-1-AP-150UL
This item is not returnable.
View return policy
Description
Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.Specifications
AMPD2 | |
Polyclonal | |
Unconjugated | |
AMPD2 | |
AMP deaminase 2, AMP deaminase isoform L, AMPD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109674, 271, 362015 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01433, Q02356, Q9DBT5 | |
Ampd2 | |
AMPD2 Fusion Protein Ag8335 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |