missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 99+%, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072835
Click to view available options
Quantity:
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
N.N.OS(O)(=O)=O
This item is not returnable. View return policy
7783-20-2
H8N2O4S
132.13
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
N.N.OS(O)(=O)=O
Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 2.5 kg
Formula Weight 132.13g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'