missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, 28 to 30%, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | NH4OH |
Concentration | 28 to 30% NH3 basis |
UN Number | 2672 |
Beilstein | 3587154 |
Density | 0.9g/mL at 25°C |
Quantity | 2.5 L |
IUPAC Name | azanium;hydroxide |
Explosion Limit | 0.27 |
Formula Weight | 35.05g/mol |
Grade | ACS Reagent |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction