missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydrogen Difluoride, 95%, Reagent grade, Honeywell™
Reagent Grade, 95%
Chemical Identifiers
CAS | 1341-49-7 |
---|---|
Molecular Formula | F2H5N |
Molecular Weight (g/mol) | 57.04 |
MDL Number | MFCD00012018 |
InChI Key | KVBCYCWRDBDGBG-UHFFFAOYSA-N |
IUPAC Name | amine dihydrofluoride |
SMILES | N.F.F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15662240
|
Honeywell Research Chemicals
224820-25G |
25 g | Poly Bottle |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
15672240
|
Honeywell Research Chemicals
224820-500G |
500 g | Poly Bottle |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
1341-49-7 | |
57.04 | |
KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
N.F.F |
F2H5N | |
MFCD00012018 | |
amine dihydrofluoride |
Specifications
125°C | |
White | |
NH4HF2 | |
UN1727 | |
N.F.F | |
57.04 | |
95% | |
Crystals or flakes | |
Ammonium Hydrogen Difluoride |
1341-49-7 | |
F2H5N | |
MFCD00012018 | |
KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
amine dihydrofluoride | |
57.04g/mol | |
Reagent | |
1.500g/cm3 at 20°C |
Safety and Handling
Missing Phrase - N15.00950417-P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H301-H314
missing translation for 'dotInformation' : 8/PGII
missing translation for 'einecsNumber' : 215-676-4
missing translation for 'rtecsNumber' : BQ9200000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title