missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bicarbonate, Reagent Grade, ≥99%, Honeywell™

Reagent Grade,≥99.0%

£14.65 - £27.46

Chemical Identifiers

CAS 1066-33-7
Molecular Formula C3H8N2O9Zn2
Molecular Weight (g/mol) 346.86
MDL Number MFCD00012138
InChI Key QOPSSYZLVIENBR-UHFFFAOYSA-J
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
SMILES [NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15647660
View Documents
Honeywell Research Chemicals
A6141-25G
25 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15657660
View Documents
Honeywell Research Chemicals
A6141-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15637660
View Documents
Honeywell Research Chemicals
A6141-1KG
1 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1066-33-7
346.86
QOPSSYZLVIENBR-UHFFFAOYSA-J
14013
C3H8N2O9Zn2
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

60°C
Colorless
C3H8N2O9Zn2
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
346.86
79.06g/mol
Ammoniacal
1.58g/cm3 at 20°C; Vapor Density: 2.7 (vs air)
1066-33-7
8 (at 20°C)
NH4HCO3
4329606
QOPSSYZLVIENBR-UHFFFAOYSA-J
azanium;hydrogen carbonate
14013
≥99%
Solid
Ammonium bicarbonate
Videos
Safety and Handling

Safety and Handling

P301 + P312 + P330

missing translation for 'rtecsNumber' : 213-911-5

Product Identifier
  • Ammonium bicarbonate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Certificates