missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, Certified AR for Analysis, Fisher Chemical™
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 g
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C2H7NO2, CAS Number-631-61-8, 500g, White, 77.08g/mol, 114 deg.C, 13066, Plastic powder jar, Solid
Specifications
Melting Point | 114°C |
Color | White |
pH | 7 |
Packaging | Plastic powder jar |
Quantity | 250 g |
Formula Weight | 77.08g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'