missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Streptavidin-HRP Conjugate
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications
£75.85 - £380.00
Specifications
For Use With (Application) | Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry |
---|---|
Includes | 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10320455
|
Cytiva
RPN1231-2ML |
2 mL |
£380.00
2mL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10535925
|
Cytiva
RPN1231-100UL |
100 mL |
£75.85
100µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- High specificity reagents
- Conjugation achieved using the periodate coupling method described by Nakane and co-workers
- Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry | |
2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent |