Learn More
Cytiva Amersham™ CyDye™ Antibody Labeling Kits
Convenient way to tag antibodies with CyDye fluorescent dyes
Brand: Cytiva PA32001
Code : GE
Additional Details : CAS Number : 497-19-8 Weight : 0.18800kg
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Conjugations are carried out under mild conditions and are complete within 1 hour
- Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
Specifications
For the conjugation of proteins with Cy2 bisfunctional dye | |
Cy2 MAb Labeling Kit | |
896.95 | |
2°C to 8°C, protected from light | |
Yellow |
Each | |
7 to 7.3 | |
Cy2 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water | |
Cy2 mono-Functional Dye |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1