Learn More
Invitrogen™ TAE (25X)
Tested for RNase and DNase contaminutesation
Brand: Invitrogen™ AM9870
Code : 50
Additional Details : Weight : 1.72000kg
Description
Invitrogen 25X TAE (High Purity Tris-Acetate-EDTA Buffer) is thoroughly tested for RNase and DNase contamination, making it ideal for use in sensitive application. Ten packets are provided, each containing the necessary powder to make 1L of 25X TAE buffer upon the addition of H2O. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Specifications:
- Appearance: White crystalline powder
- Molecular Weight: Tris-Acetate buffer component - 121.14; EDTA component - 372.3
- Compatible with agarose gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room temperature
Specifications
Running Buffers | |
25X | |
Nucleic acid gel electrophoresis & blotting | |
Ambion |
For Research Use Only. Not for use in diagnostic procedures.