missing translation for 'onlineSavingsMsg'
Learn More
Amberchrom™ 50WX8 50-100 (H), Thermo Scientific Chemicals Product Code.: 11409627

Amberchrom™ 50WX8 50-100 (H), Thermo Scientific Chemicals

Product Code. 11409627
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11409627

Brand: Thermo Scientific Alfa Aesar B22109.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dowex™ 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Quantity 2500 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 50-100 (H)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252570 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog NumberB22109.22,B22109.0E,B22109.36
Lot NumberS20K006 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog NumberB22109.22,B22109.0E,B22109.36
Lot NumberY28J055 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog NumberB22109.22,B22109.0E,B22109.36
Lot Number10240997 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberB22109.22,B22109.0E,B22109.36
Lot Number10248243 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog NumberB22109.22,B22109.0E,B22109.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberchrom™ 50WX8 50-100 (H), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.