missing translation for 'onlineSavingsMsg'
Learn More

Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%, Thermo Scientific Chemicals

£22.61 - £194.97

Chemical Identifiers

CAS 32823-06-6
MDL Number MFCD00075314
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367328
View Documents
Thermo Scientific Alfa Aesar
030715.30
250 g
£22.61
250g
Please to purchase this item. Need a web account? Register with us today!
11377328
View Documents
Thermo Scientific Alfa Aesar
030715.A1
1 kg
£56.48
1kg
Please to purchase this item. Need a web account? Register with us today!
11387328
View Documents
Thermo Scientific Alfa Aesar
030715.D9
5 x 1 kg
£194.97
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32823-06-6
MFCD00075314

Specifications

32823-06-6
Powder
Insoluble in water.
Al2O3 19.4%, P2O5 78.9%
Aluminum metaphosphate
White
MFCD00075314
263.9
2.779 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum metaphosphate

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only