missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum: ISO Safety Sign - Wear a mask

Sign compliant to ISO 7010.

Brand:  Brady™ 820218

 View more versions of this product

Product Code. 15614537

  • £39.15 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Aluminum
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 140°C
Global
Aluminum
315 mm
1.016 mm
Round
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers