missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.025mm (0.001in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

£125.63 - £294.47

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379748
SDS View Documents
Thermo Scientific Alfa Aesar
044233.GA
50 x 200 mm
£125.63
Each
Please to purchase this item. Need a web account? Register with us today!
11389748
SDS View Documents
Thermo Scientific Alfa Aesar
044233.HC
50 x 1000 mm
£294.47
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629
[Al]

Specifications

7429-90-5
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
5359268
99.999%
≈0.675g/50x200mm
(metals basis)
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Odorless
Foil
Aluminum foil, 0.025mm (0.001 in.) thick
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.