missing translation for 'onlineSavingsMsg'
Learn More
Aluminum Chloride Hexahydrate, Honeywell™ Product Code.: 15650820

Aluminum Chloride Hexahydrate, Honeywell™

Product Code. 15650820
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 15650820

Brand: Honeywell Research Chemicals 062321KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a., crystallized,≥99.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color White to Yellow
Physical Form Crystallized
Quantity 1 kg
UN Number NONH for all modes of transport
Formula Weight 241.43g/mol
Percent Purity ≥99% (AT)
Grade purum p.a.
Vapor Pressure 1mmHg (100°C)
Chemical Name or Material Aluminum Chloride Hexahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Chloride Hexahydrate, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.