missing translation for 'onlineSavingsMsg'
Learn More
Aluminum chloride, 0.5M aq. soln., Thermo Scientific™ Product Code.: 15474159

Aluminum chloride, 0.5M aq. soln., Thermo Scientific™

Product Code. 15474159
50 mL
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy

Product Code. 15474159

Brand: Thermo Scientific Alfa Aesar J61582.AD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
UN Number UN2581
IUPAC Name aluminum(3+) hexahydrate trichloride
Formula Weight 133.34
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Chemical Name or Material Aluminum chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J500 Certificate TypeCertificate of Analysis Date18/10/2023 Catalog NumberJ61582.AD
Lot NumberW15I504 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number
Lot NumberZ05E515 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberT21G500 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberG05X563 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride, 0.5M aq. soln., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.