Learn More
Alsin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561254
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FQPGLYYSGR QDPTEGDNLP ENHSGSKTPV LLSCSKLGYI SRVTAGKDSY LALVDKNIMG YIASLHELAT TERRFYSKLS D Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The function of this protein remains unknown.Specifications
Alsin | |
Polyclonal | |
Unconjugated | |
ALS2 | |
3222402C23Rik; 9430073A21Rik; Als2; ALS2, alsin Rho guanine nucleotide exchange factor; Als2cr6; Alsin; ALSJ; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; IAHSP; KIAA1563; MGC87187; mKIAA1563; PLSJ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57679 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96Q42 | |
ALS2 | |
Recombinant protein corresponding to Human Alsin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only