Learn More
Ansell™ AlphaTec™MICROGARD™2000 Model 417 Overshoes
MICROGARD™ 2000 STANDARD provides bothprotection and comfort with exceptional liquid andparticulate protection. Ideal for a wide range ofindustrial applications
£804.00 - £892.00
Specifications
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
View More Specs
Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15835352
View Documents
Ansell™
WH20B-00417-05
Large
£892.00
Pack of 200
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
15815352
View Documents
Ansell™
WH20B-00417-00
Medium
£804.00
Pack of 250
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
15825352
View Documents
Ansell™
WH20B-00417-02
Small
£824.00
Pack of 250
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
Description
Description
  • Elasticated opening
  • Bound seams
  • ESD PVC Sole
  • Size: 39-42 • 42-46 • 46-48
  • Color: White
Specifications
Specifications
Yes
Single-use
Elastic Opening
Bound
EN14126; EN1073-2; DIN32781; EN1149-5; Type 4-B; Type5-B; Type6-B
Static, Bloodborne Pathogens, Chemical-resistant, Liquid, Water
White
Non-sterile
Overshoes - ESD
Proven barrier to low concentration liquid chemicals, diluted pesticides,liquid & particulate biological hazards
250/Pk.
Shoe Covers